MilliporeSigma
  • TRPV4 regulates insulin mRNA expression and INS-1E cell death via ERK1/2 and NO-dependent mechanisms.
TRPV4 regulates insulin mRNA expression and INS-1E cell death via ERK1/2 and NO-dependent mechanisms.
Cellular signalling (2017-04-01)
M Billert, M Skrzypski, M Sassek, D Szczepankiewicz, T Wojciechowicz, S Mergler, M Z Strowski, K W Nowak
ABSTRACT
TRPV4 is a Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human TRPV4
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%