MilliporeSigma
  • Bridgehead isomer effects in bis(phosphido)-bridged diiron hexacarbonyl proton reduction electrocatalysts.
Bridgehead isomer effects in bis(phosphido)-bridged diiron hexacarbonyl proton reduction electrocatalysts.
Dalton transactions (Cambridge, England : 2003) (2017-02-22)
Ahibur Rahaman, Carolina Gimbert-Suriñach, Arne Ficks, Graham E Ball, Mohan Bhadbhade, Matti Haukka, Lee Higham, Ebbe Nordlander, Stephen B Colbran
ABSTRACT
The influence of the substitution, orientation and structure of the phosphido bridges in [Fe
Sigma-Aldrich
Phenylphosphine, ca. 10% weight in hexanes