MilliporeSigma
  • Biocompatible silver nanoparticles prepared with amino acids and a green method.
Biocompatible silver nanoparticles prepared with amino acids and a green method.
Amino acids (2016-11-30)
Ricardo Almeida de Matos, Lilia Coronato Courrol
ABSTRACT
The synthesis of nanoparticles is usually carried out by chemical reduction, which is effective but uses many toxic substances, making the process potentially harmful to the environment. Hence, as part of the search for environmentally friendly or green synthetic methods, this study aimed to produce silver nanoparticles (AgNPs) using only AgNO