MilliporeSigma
Improved electron-hole separation and migration in anatase TiO
Nanoscale (2017-03-23)
Gregor Žerjav, Muhammad Shahid Arshad, Petar Djinović, Ita Junkar, Janez Kovač, Janez Zavašnik, Albin Pintar
ABSTRACT
The as-synthesized TiO
Product Description
Ethanol solution, NIST® SRM® 2899a, nominal 25% by mass