MilliporeSigma
Increasing spinal 5-HT
Pharmacological research (2016-09-25)
Amandine Dupuis, Anne-Sophie Wattiez, Jérémy Pinguet, Damien Richard, Frédéric Libert, Maryse Chalus, Youssef Aissouni, Benoit Sion, Denis Ardid, Philippe Marin, Alain Eschalier, Christine Courteix
ABSTRACT
Antidepressants are one of the first line treatments for neuropathic pain but their use is limited by the incidence and severity of side effects of tricyclics and the weak effectiveness of selective serotonin reuptake inhibitors (SSRIs). Serotonin type 2A (5-HT