MilliporeSigma
  • Determination of monoamine neurotransmitters in zebrafish (Danio rerio) by gas chromatography coupled to mass spectrometry with a two-step derivatization.
Determination of monoamine neurotransmitters in zebrafish (Danio rerio) by gas chromatography coupled to mass spectrometry with a two-step derivatization.
Analytical and bioanalytical chemistry (2017-02-17)
Alvaro Aragon, Jessica Legradi, Ana Ballesteros-Gómez, Juliette Legler, Martin van Velzen, Jacob de Boer, Pim Leonards
ABSTRACT
A sensitive analytical method for the determination of monoamine neurotransmitters (MNTs) in zebrafish larvae was developed using gas chromatography coupled to mass spectrometry. Six MNTs were selected as target compounds for neurotoxicity testing. MNTs underwent a two-step derivatization with hexamethyldisilazane (HDMS) for O-silylation followed by N-methyl-bis-heptafluorobutyramide (MBHFBA) for N-perfluoroacylation. Derivatization conditions were optimized by an experimental design approach. Method validation showed linear calibration curves (r
Product Description
Supelco
Pirimicarb, PESTANAL®, analytical standard
Sigma-Aldrich
2-Methylheptane, 98%
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)