MilliporeSigma
WO
Physical chemistry chemical physics : PCCP (2017-01-27)
Junghyun Choi, Pitchaimuthu Sudhagar, Joo Hyun Kim, Jiseok Kwon, Jeonghyun Kim, Chiaki Terashima, Akira Fujishima, Taeseup Song, Ungyu Paik
ABSTRACT
We demonstrate the dual advantages of graded photoabsorbers in mesoporous metal oxide-based hetero interfacial photoanodes in improving photogenerated charge carrier (e
Sigma-Aldrich
Ammonium metavanadate, 99%