MilliporeSigma
SENP1 regulates PTEN stability to dictate prostate cancer development.
Oncotarget (2016-11-17)
Tasneem Bawa-Khalfe, Feng-Ming Yang, Joan Ritho, Hui-Kuan Lin, Jinke Cheng, Edward T H Yeh
ABSTRACT
SUMO protease SENP1 is elevated in multiple carcinomas including prostate cancer (PCa). SENP1 exhibits carcinogenic properties; it promotes androgen receptor-dependent and -independent cell proliferation, stabilizes HIF1α, increases VEGF, and supports angiogenesis. However, mice expressing an androgen-responsive promoter driven SENP1-transgene (SENP1-Tg) develop high-grade prostatic intraepithelial neoplasia, but not carcinoma. We now show that tumor suppressive PTEN signaling is induced in SENP1-Tg to enhance prostate epithelial cell apoptosis. SENP1 blocks SUMO1-dependent ubiquitylation and degradation of PTEN. In the absence of SENP1, SUMO1-modified PTEN is sequestered in the cytosol, where binding to ubiquitin-E3 ligase WWP2 occurs. Concurrently, WWP2 is also SUMOylated, which potentiates its interaction with PTEN. Thus, SENP1 directs ubiquitin-E3-substrate association to control PTEN stability. PTEN serves as a barrier for SENP1-mediated prostate carcinogenesis as SENP1-Tg mice develop invasive carcinomas only after PTEN reduction. Hence, SENP1 modulates multiple facets of carcinogenesis and may serve as a target specifically for aggressive PTEN-deficient PCa.