MilliporeSigma
  • Androgen receptor regulation by histone methyltransferase Suppressor of variegation 3-9 homolog 2 and Melanoma antigen-A11.
Androgen receptor regulation by histone methyltransferase Suppressor of variegation 3-9 homolog 2 and Melanoma antigen-A11.
Molecular and cellular endocrinology (2017-01-04)
Emily B Askew, Suxia Bai, Amanda B Parris, John T Minges, Elizabeth M Wilson
ABSTRACT
Androgen receptor (AR) transcriptional activity depends on interactions between the AR NH
Sigma-Aldrich
MISSION® esiRNA, targeting human SUV39H2
Pricing and availability is not currently available.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Pricing and availability is not currently available.