MilliporeSigma
  • CFTR-β-catenin interaction regulates mouse embryonic stem cell differentiation and embryonic development.
CFTR-β-catenin interaction regulates mouse embryonic stem cell differentiation and embryonic development.
Cell death and differentiation (2016-11-12)
Zhenqing Liu, Jinghui Guo, Yan Wang, Zhihui Weng, Biao Huang, Mei-Kuen Yu, Xiaohu Zhang, Ping Yuan, Hui Zhao, Wai-Yee Chan, Xiaohua Jiang, Hsiao-Chang Chan
ABSTRACT
Cystic fibrosis transmembrane conductance regulator (CFTR) is a cAMP-regulated anion channel capable of conducting both Cl- and HCO3-, mutations of which cause cystic fibrosis (CF), a common autosomal recessive disease. Although CF patients are known to have varied degree of developmental problems, the biological role of CFTR in embryonic development remains elusive. Here, we show that CFTR is functionally expressed in mouse ESCs. CFTR-/- mESCs exhibit dramatic defect in mesendoderm differentiation. In addition, CFTR physically interacts with β-catenin, defect of which leads to premature degradation of β-catenin and suppressed activation of β-catenin signaling. Furthermore, knockdown of CFTR retards the early development of Xenopus laevis with impaired mesoderm/endoderm differentiation and β-catenin signaling. Our study reveals a previously undefined role of CFTR in controlling ESC differentiation and early embryonic development via its interaction with β-catenin, and provides novel insights into the understanding of embryonic development.
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, clone MC-480, Chemicon®, from mouse
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture