MilliporeSigma
  • Effects of four different drying methods on the carotenoid composition and antioxidant capacity of dried Gac peel.
Effects of four different drying methods on the carotenoid composition and antioxidant capacity of dried Gac peel.
Journal of the science of food and agriculture (2016-07-21)
Hoang V Chuyen, Paul D Roach, John B Golding, Sophie E Parks, Minh H Nguyen
ABSTRACT
Gac fruit (Momordica cochinchinensis Spreng.) is a rich source of carotenoids for the manufacture of powder, oil and capsules for food, cosmetic and pharmaceutical uses. Currently, only the aril of the Gac fruit is processed and the peel, similar to the other components, is discarded, although it contains high level of carotenoids, which could be extracted for commercial use. In the present study, four different drying methods (hot-air, vacuum, heat pump and freeze drying), different temperatures and drying times were investigated for producing dried Gac peel suitable for carotenoid extraction. The drying methods and drying temperatures significantly affected the drying time, carotenoid content and antioxidant capacity of the dried Gac peel. Among the investigated drying methods, hot-air drying at 80 Hot-air drying at 80
Sigma-Aldrich
(S)-(−)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, 98%