MilliporeSigma
  • Discovery of 1-(3-(benzyloxy)pyridin-2-yl)-3-(2-(piperazin-1-yl)ethyl)urea: A new modulator for amyloid beta-induced mitochondrial dysfunction.
Discovery of 1-(3-(benzyloxy)pyridin-2-yl)-3-(2-(piperazin-1-yl)ethyl)urea: A new modulator for amyloid beta-induced mitochondrial dysfunction.
European journal of medicinal chemistry (2017-02-06)
Ahmed Elkamhawy, Jung-Eun Park, Ahmed H E Hassan, Hyunhwa Ra, Ae Nim Pae, Jiyoun Lee, Beoung-Geon Park, Bongjin Moon, Hyun-Mee Park, Eun Joo Roh
ABSTRACT
Herein, we report a new series of aliphatic substituted pyridyl-urea small molecules synthesized as potential modulators for amyloid beta (Aβ) induced mitochondrial dysfunction. Their blocking activities against Aβ-induced mitochondrial permeability transition pore (mPTP) opening were evaluated by JC-1 assay which measures the change of mitochondrial membrane potential (ΔΨm). The inhibitory activity of sixteen compounds against Aβ-induced mPTP opening was superior or almost similar to that of the standard Cyclosporin A (CsA). Among them, 1-(3-(benzyloxy)pyridin-2-yl)-3-(2-(piperazin-1-yl)ethyl)urea (5x) effectively maintained mitochondrial function and cell viabilities on ATP assay, MTT assay, and ROS assay. Using CDocker algorithm, a molecular docking model presented a plausible binding mode for 5x with cyclophilin D (CypD) receptor as a major component of mPTP. Moreover, hERG and BBB-PAMPA assays presented safe cardiotoxicity and high CNS bioavailability profiles for 5x. Taken as a whole, this report presents compound 5x as a new nonpeptidyl mPTP blocker may hold a promise for further development of Alzheimer's disease (AD) therapeutics.
Sigma-Aldrich
2-Amino-3-benzyloxypyridine, 99%
Sigma-Aldrich
JC-1, solid