MilliporeSigma
  • Superficially porous particles with 1000Å pores for large biomolecule high performance liquid chromatography and polymer size exclusion chromatography.
Superficially porous particles with 1000Å pores for large biomolecule high performance liquid chromatography and polymer size exclusion chromatography.
Journal of chromatography. A (2017-02-19)
Brian M Wagner, Stephanie A Schuster, Barry E Boyes, Taylor J Shields, William L Miles, Mark J Haynes, Robert E Moran, Joseph J Kirkland, Mark R Schure
ABSTRACT
To facilitate mass transport and column efficiency, solutes must have free access to particle pores to facilitate interactions with the stationary phase. To ensure this feature, particles should be used for HPLC separations which have pores sufficiently large to accommodate the solute without restricted diffusion. This paper describes the design and properties of superficially porous (also called Fused-Core
Sigma-Aldrich
Difluoroacetic acid, 98%