MilliporeSigma
  • MicroRNA-124 regulates the radiosensitivity of non-small cell lung cancer cells by targeting TXNRD1.
MicroRNA-124 regulates the radiosensitivity of non-small cell lung cancer cells by targeting TXNRD1.
Oncology letters (2017-04-30)
Chuncheng Hao, Xiangying Xu, Jia Ma, Jun Xia, Bingbing Dai, Lili Liu, Yuyan Ma
ABSTRACT
Radiation treatment remains one of the major modalities in the treatment of lung cancer. Although the majority of patients initially respond to treatment with radiation, resistance inevitably develops and leads to treatment failure. Therefore, the identification of the underlying molecular mechanisms of radiation resistance may facilitate the development of novel approaches for overcoming resistance, and enhance the efficacy of treatment with radiation in lung and other types of cancer. In the present study we established three radiation-resistant sub-cell lines derived from the radiation-sensitive lung cancer cell line HCC827. Using a polymerase chain reaction microRNA (miRNA) array, multiple miRNAs were identified to be markedly downregulated in radiation-resistant cells, including miRNA (miR)-124, miR-191 and miR-205. It was observed that overexpression of miR-124 sensitized the resistant cells to treatment with radiation and that thioredoxin reductase 1 (TXNRD1) is a novel target of miR-124. Furthermore, it was demonstrated that knockdown of TXNRD1 using small interfering RNA increased the basal level of reactive oxygen species and sensitized the cells to radiation treatment. The results of the present study demonstrated that multiple miRNAs are downregulated in radiation-resistant lung cancer cells and that downregulation of miR-124 mediates radiation resistance through the targeting of TXNRD1 mRNA expression. The present study revealed a novel molecular mechanism of miRNA-mediated radiation resistance and identified miR-124-regulated TXNRD1 as a novel therapeutic target for overcoming radiation resistance in the treatment of lung cancer.
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Anti-beta-Actin Antibody, clone 4C2, clone 4C2, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human TXNRD1
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%