MilliporeSigma
  • Boronic acid catalyzed ene carbocyclization of acetylenic dicarbonyl compounds.
Boronic acid catalyzed ene carbocyclization of acetylenic dicarbonyl compounds.
Chemical communications (Cambridge, England) (2010-03-18)
Meiling Li, Ting Yang, Darren J Dixon
ABSTRACT
The discovery and development of an efficient ene carbocyclization of 1,3-dicarbonyl compounds bearing pendent terminal alkyne substituents under 3-nitrobenzeneboronic acid catalysis is described. The reaction is efficient, easy to perform and general to a wide range of ketoester substrates.
Sigma-Aldrich
3-Nitrophenylboronic acid, ≥97%