MilliporeSigma
  • Synthesis of phosphorus-doped graphene and its multifunctional applications for oxygen reduction reaction and lithium ion batteries.
Synthesis of phosphorus-doped graphene and its multifunctional applications for oxygen reduction reaction and lithium ion batteries.
Advanced materials (Deerfield Beach, Fla.) (2013-07-19)
Chenzhen Zhang, Nasir Mahmood, Han Yin, Fei Liu, Yanglong Hou
ABSTRACT
We develop a simple and economical thermal annealing method for the synthesis of phosphorus-doped graphene, which exhibits remarkable electrocatalytic activity towards the oxygen reduction reaction and enhances the electrochemical performance as an anode material for lithium ion batteries. The experimental results suggest the significant role of phosphorus atoms in graphene.
Sigma-Aldrich
Phosphorus-doped graphene, avg. no. of layers, 1 ‑ 5