MilliporeSigma
  • Multi-residue determination of micropollutants in Phragmites australis from constructed wetlands using microwave assisted extraction and ultra-high-performance liquid chromatography tandem mass spectrometry.
Multi-residue determination of micropollutants in Phragmites australis from constructed wetlands using microwave assisted extraction and ultra-high-performance liquid chromatography tandem mass spectrometry.
Analytica chimica acta (2017-02-06)
Bruce Petrie, Benjamin D Smith, Jane Youdan, Ruth Barden, Barbara Kasprzyk-Hordern
ABSTRACT
In constructed wetlands micropollutants can be removed from water by phytoremediation. However, micropollutant uptake and metabolism by plants here is poorly understood due to the lack of good analytical approaches. Reported herein is the first methodology developed and validated for the multi-residue determination of 81 micropollutants (pharmaceuticals, personal care products and illicit drugs) in the emergent macrophyte Phragmites australis. The method involved extraction by microwave accelerated extraction (MAE), clean-up using off-line solid phase extraction and analysis by ultra-high-performance liquid chromatography tandem mass spectrometry. Development of the MAE method found the influence of studied variables on micropollutant recovery to be: extraction temperature > sample mass > solvent composition. Validation of the developed extraction protocol revealed method recoveries were in the range 80-120% for the majority of micropollutants. Method quantitation limits (MQLs) were generally <5 ng g
Product Description
Supelco
Ketoprofen-d3, VETRANAL®, analytical standard