MilliporeSigma
  • Targeted osteosarcoma chemotherapy using RGD peptide-installed doxorubicin-loaded biodegradable polymeric micelle.
Targeted osteosarcoma chemotherapy using RGD peptide-installed doxorubicin-loaded biodegradable polymeric micelle.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-12-09)
Zhenhua Fang, Yanpeng Sun, Hong Xiao, Peng Li, Ming Liu, Fan Ding, Wusheng Kan, Runsheng Miao
ABSTRACT
Osteosarcoma is the most common primary malignant bone tumor in the pediatric age group, and chemotherapy directed by targeted nanoparticulate drug delivery system represents a promising approach for osteosarcoma treatment recently. Here, we designed and developed a novel DOX-loaded targeted polymeric micelle self-assembled from RGD-terminated poly(ethylene glycol)-block-poly (trimethylene carbonate) (RGD-PEG-PTMC) amphiphilic biodegradable block copolymer, for high-efficiency targeted chemotherapy of osteosarcoma. Notably, the RGD-installed DOX-loaded biodegradable polymeric micelle (RGD-DOX-PM) with drug loading efficiency of 57%-73% displayed a narrow distribution (PDI=0.05-0.12) with average sizes ranging from 46 to 73nm depending on the DOX loading content. The release amount of DOX from RGD-DOX-PM achieved 63% within 60h under physiological condition. Interestingly, MTT assays in MG-63 and MNNG/HOS osteosarcoma cells exhibited that half-maximal inhibitory concentration (IC
Sigma-Aldrich
Diphenyl phosphite