MilliporeSigma
  • Development of a disposable and low-cost electrochemical sensor for dopamine detection based on poly(pyrrole-3-carboxylic acid)-modified electrochemically over-oxidized pencil graphite electrode.
Development of a disposable and low-cost electrochemical sensor for dopamine detection based on poly(pyrrole-3-carboxylic acid)-modified electrochemically over-oxidized pencil graphite electrode.
Talanta (2017-02-06)
Ali Özcan, Salih İlkbaş, Ayça Atılır Özcan
ABSTRACT
In this study, preparation of a single-use electrochemical sensor for the selective and sensitive determination of dopamine (DOP) was investigated by electrochemical polymerization of pyrrole-3-carboxylic acid on electrochemically over-oxidized pencil graphite electrode (p(P3CA)/EOPGE). Cyclic voltammetry measurements of Fe(CN)
Sigma-Aldrich
Ruthenium(III) chloride, Ru content 45-55%
Sigma-Aldrich
Pyrrole-3-carboxylic acid, ≥96%