MilliporeSigma
  • Heterolytic OO bond cleavage: Functional role of Glu113 during bis-Fe(IV) formation in MauG.
Heterolytic OO bond cleavage: Functional role of Glu113 during bis-Fe(IV) formation in MauG.
Journal of inorganic biochemistry (2016-12-03)
Jiafeng Geng, Lu Huo, Aimin Liu
ABSTRACT
The diheme enzyme MauG utilizes H
Sigma-Aldrich
2-Phenyl-2-propanol, 97%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%