MilliporeSigma
  • A New Caged-Glutamine Derivative as a Tool To Control the Assembly of Glutamine-Containing Amyloidogenic Peptides.
A New Caged-Glutamine Derivative as a Tool To Control the Assembly of Glutamine-Containing Amyloidogenic Peptides.
Chembiochem : a European journal of chemical biology (2016-10-28)
Loay Awad, Nino Jejelava, Ritwik Burai, Hilal A Lashuel
ABSTRACT
We present the design, synthesis, and characterization of a novel photocaged glutamine derivative (modified on the side chain of glutamine), and describe its use in enhancing peptide stability and solubility. Our results demonstrate that this approach can be used to develop molecular switches to control the folding and β-sheet formation of amyloidogenic peptides.
Sigma-Aldrich
Fmoc-Glu(OtBu)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Fmoc-Leu-OH, ≥97.0%
Sigma-Aldrich
Fmoc-Tyr(tBu)-OH, ≥98.0% (HPLC)