Merck
Catfish rhamnose-binding lectin induces G
Glycoconjugate journal (2016-11-01)
Shigeki Sugawara, Changhun Im, Tasuku Kawano, Takeo Tatsuta, Yasuhiro Koide, Daiki Yamamoto, Yasuhiro Ozeki, Kazuo Nitta, Masahiro Hosono
ABSTRACT
Silurus asotus egg lectin (SAL), an α-galactoside-binding protein isolated from the eggs of catfish, is a member of the rhamnose-binding lectin family that binds to Gb3 glycan (Galα1-4Galβ1-4Glc). We have previously demonstrated that SAL reduces the proliferation of Gb3-expressing Burkitt's lymphoma Raji cells and confirm here that it does not reduce their viability, indicating that unlike other lectins, it is not cytotoxic. The aim of this study was to determine the signal transduction mechanism(s) underlying this novel SAL/Gb3 binding-mediated effect profile. SAL/Gb3 interaction arrested the cell cycle through increasing the G
Sigma-Aldrich
MISSION® esiRNA, targeting human A4GALT