MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Systematic Identification of Oncogenic EGFR Interaction Partners.
Journal of molecular biology (2016-12-14)
Julia Petschnigg, Max Kotlyar, Louise Blair, Igor Jurisica, Igor Stagljar, Robin Ketteler
ABSTRACT
The epidermal growth factor receptor (EGFR) is a receptor tyrosine kinase (TK) that-once activated upon ligand binding-leads to receptor dimerization, recruitment of protein complexes, and activation of multiple signaling cascades. The EGFR is frequently overexpressed or mutated in various cancers leading to aberrant signaling and tumor growth. Hence, identification of interaction partners that bind to mutated EGFR can help identify novel targets for drug discovery. Here, we used a systematic approach to identify novel proteins that are involved in cancerous EGFR signaling. Using a combination of high-content imaging and a mammalian membrane two-hybrid protein-protein interaction method, we identified eight novel interaction partners of EGFR, of which half strongly interacted with oncogenic, hyperactive EGFR variants. One of these, transforming acidic coiled-coil proteins (TACC) 3, stabilizes EGFR on the cell surface, which results in an increase in downstream signaling via the mitogen-activated protein kinase and AKT pathway. Depletion of TACC3 from cells using small hairpin RNA (shRNA) knockdown or small-molecule targeting reduced mitogenic signaling in non-small cell lung cancer cell lines, suggesting that targeting TACC3 has potential as a new therapeutic approach for non-small cell lung cancer.