MilliporeSigma
  • Conductive porous vanadium nitride/graphene composite as chemical anchor of polysulfides for lithium-sulfur batteries.
Conductive porous vanadium nitride/graphene composite as chemical anchor of polysulfides for lithium-sulfur batteries.
Nature communications (2017-03-04)
Zhenhua Sun, Jingqi Zhang, Lichang Yin, Guangjian Hu, Ruopian Fang, Hui-Ming Cheng, Feng Li
ABSTRACT
Although the rechargeable lithium-sulfur battery is an advanced energy storage system, its practical implementation has been impeded by many issues, in particular the shuttle effect causing rapid capacity fade and low Coulombic efficiency. Herein, we report a conductive porous vanadium nitride nanoribbon/graphene composite accommodating the catholyte as the cathode of a lithium-sulfur battery. The vanadium nitride/graphene composite provides strong anchoring for polysulfides and fast polysulfide conversion. The anchoring effect of vanadium nitride is confirmed by experimental and theoretical results. Owing to the high conductivity of vanadium nitride, the composite cathode exhibits lower polarization and faster redox reaction kinetics than a reduced graphene oxide cathode, showing good rate and cycling performances. The initial capacity reaches 1,471 mAh g
Sigma-Aldrich
1,2-Dimethoxyethane, anhydrous, 99.5%, inhibitor-free