MilliporeSigma
  • Assessment of myocardial injury after reperfused infarction by T1ρ cardiovascular magnetic resonance.
Assessment of myocardial injury after reperfused infarction by T1ρ cardiovascular magnetic resonance.
Journal of cardiovascular magnetic resonance : official journal of the Society for Cardiovascular Magnetic Resonance (2017-02-16)
Rutger H Stoffers, Marie Madden, Mohammed Shahid, Francisco Contijoch, Joseph Solomon, James J Pilla, Joseph H Gorman, Robert C Gorman, Walter R T Witschey
ABSTRACT
The evolution of T1ρ and of other endogenous contrast methods (T2, T1) in the first month after reperfused myocardial infarction (MI) is uncertain. We conducted a study of reperfused MI in pigs to serially monitor T1ρ, T2 and T1 relaxation, scar size and transmurality at 1 and 4 weeks post-MI. Ten Yorkshire swine underwent 90 min of occlusion of the circumflex artery and reperfusion. T1ρ, T2 and native T1 maps and late gadolinium enhanced (LGE) cardiovascular magnetic resonance (CMR) data were collected at 1 week (n = 10) and 4 weeks (n = 5). Semi-automatic FWHM (full width half maximum) thresholding was used to assess scar size and transmurality and compared to histology. Relaxation times and contrast-to-noise ratio were compared in healthy and remote myocardium at 1 and 4 weeks. Linear regression and Bland-Altman was performed to compare infarct size and transmurality. Relaxation time differences between infarcted and remote myocardial tissue were ∆T1 (infarct-remote) = 421.3 ± 108.8 (1 week) and 480.0 ± 33.2 ms (4 week), ∆T1ρ = 68.1 ± 11.6 and 74.3 ± 14.2, and ∆T2 = 51.0 ± 10.1 and 59.2 ± 11.4 ms. Contrast-to-noise ratio was CNR T1ρ was highly correlated with alterations left ventricle (LV) pathology at 1 and 4 weeks post-MI and therefore it may be a useful method endogenous contrast imaging of infarction.
Product Description
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC