MilliporeSigma
  • The correlations of the electronic structure and film growth of 2,7-diocty[1]benzothieno[3,2-b]benzothiophene (C8-BTBT) on SiO
The correlations of the electronic structure and film growth of 2,7-diocty[1]benzothieno[3,2-b]benzothiophene (C8-BTBT) on SiO
Physical chemistry chemical physics : PCCP (2016-12-21)
Lu Lyu, Dongmei Niu, Haipeng Xie, Yuan Zhao, Ningtong Cao, Hong Zhang, Yuhe Zhang, Peng Liu, Yongli Gao
ABSTRACT
Combining ultraviolet photoemission spectroscopy (UPS), X-ray photoemission spectroscopy (XPS), atomic force microscopy (AFM) and small angle X-ray diffraction (SAXD) measurements, we perform a systematic investigation on the correlations of the electronic structure, film growth and molecular orientation of 2,7-diocty[1]benzothieno[3,2-b]benzothiophene (C8-BTBT) on silicon oxide (SiO