MilliporeSigma
  • Linking loggerhead locations: using multiple methods to determine the origin of sea turtles in feeding grounds.
Linking loggerhead locations: using multiple methods to determine the origin of sea turtles in feeding grounds.
Marine biology (2017-01-31)
ALan F Rees, Carlos Carreras, Annette C Broderick, Dimitris Margaritoulis, Thomas B Stringell, Brendan J Godley
ABSTRACT
Many marine megavertebrate taxa, including sea turtles, disperse widely from their hatching or birthing locations but display natal homing as adults. We used flipper tagging, satellite tracking and genetics to identify the origin of loggerhead turtles living in Amvrakikos Gulf, Greece. This location has been identified as hosting regionally important numbers of large-juvenile to adult sized turtles that display long-term residency and/or association to the area, and also presents a male biased sex ratio for adults. A total of 20 individuals were linked to nesting areas in Greece through flipper tagging and satellite telemetry, with the majority (16) associated with Zakynthos Island. One additional female was tracked from Amvrakikos Gulf to Turkey where she likely nested. Mitochondrial DNA mixed stock analyses of turtles captured in Amvrakikos Gulf (
Sigma-Aldrich
Exo 1, ≥98% (HPLC), solid