MilliporeSigma
  • Grafting and Paladin Pic-21 for Nematode and Weed Management in Vegetable Production.
Grafting and Paladin Pic-21 for Nematode and Weed Management in Vegetable Production.
Journal of nematology (2017-02-06)
Nancy Kokalis-Burelle, David M Butler, Jason C Hong, Michael G Bausher, Greg McCollum, Erin N Rosskopf
ABSTRACT
Two years of field trials conducted in a
Product Description
Supelco
Metolachlor, predominantly S-Metolachlor, PESTANAL®, analytical standard