MilliporeSigma
  • Development of chronic allergic responses by dampening Bcl6-mediated suppressor activity in memory T helper 2 cells.
Development of chronic allergic responses by dampening Bcl6-mediated suppressor activity in memory T helper 2 cells.
Proceedings of the National Academy of Sciences of the United States of America (2017-01-18)
Takashi Ogasawara, Masahiko Hatano, Hisae Satake, Jun Ikari, Toshibumi Taniguchi, Nobuhide Tsuruoka, Haruko Watanabe-Takano, Lisa Fujimura, Akemi Sakamoto, Hirokuni Hirata, Kumiya Sugiyama, Yasutsugu Fukushima, Susumu Nakae, Kenji Matsumoto, Hirohisa Saito, Takeshi Fukuda, Kazuhiro Kurasawa, Koichiro Tatsumi, Takeshi Tokuhisa, Masafumi Arima
ABSTRACT
Mice deficient in the transcriptional repressor B-cell CLL/lymphoma 6 (Bcl6) exhibit similar T helper 2 (T
Sigma-Aldrich
Human IL-33 ELISA Kit, for serum, plasma, cell culture supernatant and urine