MilliporeSigma
A simple capacitive method to evaluate ethanol fuel samples.
Scientific reports (2017-02-28)
Tatiana P Vello, Rafael F de Oliveira, Gustavo O Silva, Davi H S de Camargo, Carlos C B Bufon
ABSTRACT
Ethanol is a biofuel used worldwide. However, the presence of excessive water either during the distillation process or by fraudulent adulteration is a major concern in the use of ethanol fuel. High water levels may cause engine malfunction, in addition to being considered illegal. Here, we describe the development of a simple, fast and accurate platform based on nanostructured sensors to evaluate ethanol samples. The device fabrication is facile, based on standard microfabrication and thin-film deposition methods. The sensor operation relies on capacitance measurements employing a parallel plate capacitor containing a conformational aluminum oxide (Al