MilliporeSigma
  • Unexpected role for p19INK4d in posttranscriptional regulation of GATA1 and modulation of human terminal erythropoiesis.
Unexpected role for p19INK4d in posttranscriptional regulation of GATA1 and modulation of human terminal erythropoiesis.
Blood (2016-11-24)
Xu Han, Jieying Zhang, Yuanliang Peng, Minyuan Peng, Xiao Chen, Huiyong Chen, Jianhui Song, Xiao Hu, Mao Ye, Jianglin Li, Vijay G Sankaran, Christopher D Hillyer, Narla Mohandas, Xiuli An, Jing Liu
ABSTRACT
Terminal erythroid differentiation is tightly coordinated with cell-cycle exit, which is regulated by cyclins, cyclin-dependent kinases, and cyclin-dependent kinase inhibitors (CDKI), yet their roles in erythropoiesis remain to be fully defined. We show here that p19
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human CDKN2D