MilliporeSigma
  • CD151, a laminin receptor showing increased expression in asthmatic patients, contributes to airway hyperresponsiveness through calcium signaling.
CD151, a laminin receptor showing increased expression in asthmatic patients, contributes to airway hyperresponsiveness through calcium signaling.
The Journal of allergy and clinical immunology (2016-05-29)
Yongkang Qiao, John Kit Chung Tam, Sheryl S L Tan, Yee Kit Tai, Chin Yein Chin, Alastair G Stewart, Leonie Ashman, Kiyotoshi Sekiguchi, Shenna Y Langenbach, Gerald Stelmack, Andrew J Halayko, Thai Tran
ABSTRACT
Airway smooth muscle (ASM) contraction underpins airway constriction; however, underlying mechanisms for airway hyperresponsiveness (AHR) remain incompletely defined. CD151, a 4-transmembrane glycoprotein that associates with laminin-binding integrins, is highly expressed in the human lung. The role of CD151 in ASM function and its relationship to asthma have yet to be elucidated. We sought to ascertain whether CD151 expression is clinically relevant to asthma and whether CD151 expression affects AHR. Using immunohistochemical analysis, we determined the expression of CD151 in human bronchial biopsy specimens from patients with varying asthma severities and studied the mechanism of action of CD151 in the regulation of ASM contraction and bronchial caliber in vitro, ex vivo, and in vivo. The number of CD151 We identify a role for CD151 in human ASM contraction. We implicate CD151 as a determinant of AHR in vivo, likely through regulation of GPCR-induced calcium and PKC signaling. These observations have significant implications in understanding the mechanism for AHR and the efficacy of new and emerging therapeutics.
Sigma-Aldrich
MISSION® esiRNA, targeting human CD151