MilliporeSigma
  • Oral administration with diosgenin enhances the induction of intestinal T helper 1-like regulatory T cells in a murine model of food allergy.
Oral administration with diosgenin enhances the induction of intestinal T helper 1-like regulatory T cells in a murine model of food allergy.
International immunopharmacology (2016-11-26)
Chung-Hsiung Huang, Chia-Chi Wang, Yu-Chin Lin, Masatoshi Hori, Tong-Rong Jan
ABSTRACT
Although the development of T helper (Th)1-like regulatory T (Treg) cells under Th1 inflammatory conditions has been reported, the role of Th1-like Treg cells in Th2 allergic responses remains mostly unclear. We previously demonstrated that diosgenin, the major sapogenin contained in the Chinese yam, attenuated food allergy and augmented Th1 and Treg immune responses. In this study, we hypothesized that diosgenin may enhance the induction of Th1-like Treg cells in the gut of mice with food allergy. Ovalbumin (OVA)-sensitized BALB/c mice were gavaged daily with diosgenin and received repeatedly intragastric ovalbumin challenges to induce intestinal allergic responses. The induction of Foxp3
Sigma-Aldrich
Diosgenin, ≥93%