MilliporeSigma
  • Extraction of arabinoxylans from wheat bran using hydrothermal processes assisted by heterogeneous catalysts.
Extraction of arabinoxylans from wheat bran using hydrothermal processes assisted by heterogeneous catalysts.
Carbohydrate polymers (2017-01-25)
Nuria Sánchez-Bastardo, Alberto Romero, Esther Alonso
ABSTRACT
The extraction/hydrolysis process of arabinoxylans from destarched wheat bran was studied in this work using different mesoporous silica supports and the corresponding RuCl