- Generation of α-1,3-galactosyltransferase knocked-out transgenic cloned pigs with knocked-in five human genes.
Generation of α-1,3-galactosyltransferase knocked-out transgenic cloned pigs with knocked-in five human genes.
Transgenic research (2016-08-25)
Dae-Jin Kwon, Dong-Hwan Kim, In-Sul Hwang, Dong-Ern Kim, Hyung-Joo Kim, Jang-Seong Kim, Kichoon Lee, Gi-Sun Im, Jeong-Woong Lee, Seongsoo Hwang
PMID27554374
ABSTRACT
Recent progress in genetic manipulation of pigs designated for xenotransplantation ha6s shown considerable promise on xenograft survival in primates. However, genetic modification of multiple genes in donor pigs by knock-out and knock-in technologies, aiming to enhance immunological tolerance against transplanted organs in the recipients, has not been evaluated for health issues of donor pigs. We produced transgenic Massachusetts General Hospital piglets by knocking-out the α-1,3-galactosyltransferase (GT) gene and by simultaneously knocking-in an expression cassette containing five different human genes including, DAF, CD39, TFPI, C1 inhibitor (C1-INH), and TNFAIP3 (A20) [GT
Sigma-Aldrich
Anti-ENTPD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TFPI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution