MilliporeSigma
  • The function of cancer-shed gangliosides in macrophage phenotype: involvement with angiogenesis.
The function of cancer-shed gangliosides in macrophage phenotype: involvement with angiogenesis.
Oncotarget (2016-12-30)
Tae-Wook Chung, Hee-Jung Choi, Mi-Ju Park, Hee-Jin Choi, Syng-Ook Lee, Keuk-Jun Kim, Cheorl-Ho Kim, Changwan Hong, Kyun-Ha Kim, Myungsoo Joo, Ki-Tae Ha
ABSTRACT
Tumor-derived gangliosides in the tumor microenvironment are involved in the malignant progression of cancer. However, the molecular mechanisms underlying the effects of gangliosides shed from tumors on macrophage phenotype remain unknown. Here, we showed that ganglioside GM1 highly induced the activity and expression of arginase-1 (Arg-1), a major M2 macrophage marker, compared to various gangliosides in bone marrow-derived macrophages (BMDM), peritoneal macrophages and Raw264.7 macrophage cells. We found that GM1 bound to macrophage mannose receptor (MMR/CD206) and common gamma chain (γc). In addition, GM1 increased Arg-1 expression through CD206 and γc-mediated activation of Janus kinase 3 (JAK3) and signal transducer and activator of transcription- 6 (STAT-6). Interestingly, GM1-stimulated macrophages secreted monocyte chemoattractant protein-1 (MCP-1/CCL2) through a CD206/γc/STAT6-mediated signaling pathway and induced angiogenesis. Moreover, the angiogenic effect of GM1-treated macrophages was diminished by RS102895, an MCP-1 receptor (CCR2) antagonist. From these results we suggest that tumor-shed ganglioside is a secretory factor regulating the phenotype of macrophages and consequently enhancing angiogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human MRC1
Sigma-Aldrich
N-Acetyl-D-galactosamine, ≥98% (HPLC)