MilliporeSigma
  • Amplicon-Based RNA Interference Targeting V2 Gene of Cotton Leaf Curl Kokhran Virus-Burewala Strain Can Provide Resistance in Transgenic Cotton Plants.
Amplicon-Based RNA Interference Targeting V2 Gene of Cotton Leaf Curl Kokhran Virus-Burewala Strain Can Provide Resistance in Transgenic Cotton Plants.
Molecular biotechnology (2016-10-21)
Aneela Yasmeen, Sarfraz Kiani, Afshan Butt, Abdul Qayyum Rao, Faheem Akram, Aftab Ahmad, Idrees Ahmad Nasir, Tayyab Husnain, Shahid Mansoor, Imran Amin, Shaheen Aftab, Muhammad Zubair, Muhammad Nouman Tahir, Sohail Akhtar, Jodi Scheffler, Brian Scheffler
ABSTRACT
The conserved coat or V2 gene of begomoviruses is responsible for viral movement in the plant cells. RNAi technology was used to silence V2 gene for resistance against these viruses in transgenic plants. The transformation of the RNAi-based gene construct targeting V2 gene of CLCuKoV-Bur, cloned under 35S promoter, was done in two elite cotton varieties MNH-786 and VH-289 using shoot apex cut method of gene transformation. The transformation efficiency was found to be 3.75 and 2.88Ā % in MNH-786 and VH-289, respectively. Confirmation of successful transformation was done through PCR in T
Sigma-Aldrich
SIGMAFASTā„¢ BCIPĀ®/NBT, tablet
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent