MilliporeSigma
  • Cytotoxic and antimicrobial effects of indium(iii) complexes with 2-acetylpyridine-derived thiosemicarbazones.
Cytotoxic and antimicrobial effects of indium(iii) complexes with 2-acetylpyridine-derived thiosemicarbazones.
Dalton transactions (Cambridge, England : 2003) (2016-12-24)
Alexandre A Oliveira, Gabriele M C Perdigão, Luana E Rodrigues, Jeferson G da Silva, Elaine M Souza-Fagundes, Jacqueline A Takahashi, Willian R Rocha, Heloisa Beraldo
ABSTRACT
Complexes [In(2Ac4oClPh)Cl