MilliporeSigma
  • T-cadherin deficiency increases vascular vulnerability in T2DM through impaired NO bioactivity.
T-cadherin deficiency increases vascular vulnerability in T2DM through impaired NO bioactivity.
Cardiovascular diabetology (2017-01-21)
Han Wang, Ling Tao, Anastasia Ambrosio, Wenjun Yan, Ross Summer, Wayne Bond Lau, Yajing Wang, Xinliang Ma
ABSTRACT
Endothelial dysfunction plays a critical role in the development of type 2 diabetes (T2DM). T-cadherin (T-cad) has gained recognition as a regulator of endothelial cell (EC) function. The present study examined whether T-cad deficiency increases vascular vulnerability in T2DM. Vascular segments were isolated from WT or T-cad knockout mice. Endothelial function, total NO accumulation, and the expression of T-cad related proteins were determined. Ach and acidified NaNO2 induced similar vasorelaxation in WT groups. T-cad KO mice exhibited normal response to acidified NaNO2, but manifested markedly reduced response to Ach. NO accumulation was also decreased in T-cad KO group. T-cad expression was reduced in WT mice fed 8 weeks of high fat diet (HFD). Furthermore, exacerbated reduction of vasorelaxation was observed in T-cad KO mice fed 8 weeks of HFD. In the current study, we provide the first in vivo evidence that T-cadherin deficiency causes endothelial dysfunction in T2DM vascular segments, suggesting the involvement of T-cad deficiency in T2DM pathogenesis.
Sigma-Aldrich
Akt1/2 kinase inhibitor, ≥98% (HPLC)
Sigma-Aldrich
Anti-Nitrotyrosine Antibody, clone 1A6, clone 1A6, Upstate®, from mouse