MilliporeSigma
  • Increasing levels of interleukin (IL)-1Ra and IL-6 during the first 2 days of hospitalization in unstable angina are associated with increased risk of in-hospital coronary events.
Increasing levels of interleukin (IL)-1Ra and IL-6 during the first 2 days of hospitalization in unstable angina are associated with increased risk of in-hospital coronary events.
Circulation (1999-04-27)
L M Biasucci, G Liuzzo, G Fantuzzi, G Caligiuri, A G Rebuzzi, F Ginnetti, C A Dinarello, A Maseri
ABSTRACT
A growing body of evidence suggests a role for inflammation in acute coronary syndromes. The aim of this study was to assess the role of proinflammatory cytokines, their time course, and their association with prognosis in unstable angina. We studied 43 patients aged 62+/-8 years admitted to our coronary care unit for Braunwald class IIIB unstable angina. In each patient, serum levels of interleukin-1 receptor antagonist (IL-1Ra), interleukin-6 (IL-6) (which represent sensitive markers of biologically active IL-1beta and tumor necrosis factor-alpha levels, respectively), and troponin T were measured at entry and 48 hours after admission. Troponin T-positive patients were excluded. Patients were divided a posteriori into 2 groups according to their in-hospital outcome: group 1 comprised 17 patients with an uneventful course, and group 2 comprised 26 patients with a complicated in-hospital course. In group 1, mean IL-1Ra decreased at 48 hours by 12%, and IL-6 diminished at 48 hours by 13%. In group 2, IL-1Ra and IL-6 entry levels were higher than in group 1 and increased respectively by 37% and 57% at 48 hours (P<0.01). These findings indicate that although they receive the same medical therapy as patients who do not experience an in-hospital event, patients with unstable angina and with complicated in-hospital courses have higher cytokine levels on admission. A fall in IL-1Ra and IL-6 48 hours after admission was associated with an uneventful course and their increase with a complicated hospital course. These findings may suggest novel therapeutic approaches to patients with unstable angina.
Sigma-Aldrich
Graphene ink in water, inkjet printable
Sigma-Aldrich
SILuRich Anti-IL6 MAb, biotin conjugated, clone IL6-4G1, purified from hybridoma cell culture