MilliporeSigma
  • The presence of a single-nucleotide polymorphism in the BDNF gene affects the rate of locomotor adaptation after stroke.
The presence of a single-nucleotide polymorphism in the BDNF gene affects the rate of locomotor adaptation after stroke.
Experimental brain research (2015-10-22)
Erin E Helm, Christine M Tyrell, Ryan T Pohlig, Lucas D Brady, Darcy S Reisman
ABSTRACT
Induction of neural plasticity through motor learning has been demonstrated in animals and humans. Brain-derived neurotrophic factor (BDNF), a member of the neurotrophin family of growth factors, is thought to play an integral role in modulation of central nervous system plasticity during learning and motor skill recovery. Thirty percent of humans possess a single-nucleotide polymorphism on the BDNF gene (Val66Met), which has been linked to decreased activity-dependent release of BDNF. Presence of the polymorphism has been associated with altered cortical activation, short-term plasticity and altered skill acquisition, and learning in healthy humans. The impact of the Val66Met polymorphism on motor learning post-stroke has not been explored. The purpose of this study was to examine the impact of the Val66Met polymorphism in learning of a novel locomotor task in subjects with chronic stroke. It was hypothesized that subjects with the polymorphism would have an altered rate and magnitude of adaptation to a novel locomotor walking paradigm (the split-belt treadmill), compared to those without the polymorphism. The rate of adaptation was evaluated as the reduction in gait asymmetry during the first 30 (early adaptation) and last 100 (late adaptation) strides. Twenty-seven individuals with chronic stroke participated in a single session of split-belt treadmill walking and tested for the polymorphism. Step length and limb phase were measured to assess adaptation of spatial and temporal parameters of walking. The rate of adaptation of step length asymmetry differed significantly between those with and without the polymorphism, while the amount of total adaptation did not. These results suggest that chronic stroke survivors, regardless of presence or absence of the polymorphism, are able to adapt their walking pattern over a period of trial-and-error practice; however, the presence of the polymorphism influences the rate at which this is achieved.
Sigma-Aldrich
BDNF human, Carrier free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), suitable for cell culture
Sigma-Aldrich
Human BDNF ELISA Kit, for serum, plasma, cell culture supernatant and urine