MilliporeSigma
  • Presentation and detection of azide functionality in bacterial cell surface proteins.
Presentation and detection of azide functionality in bacterial cell surface proteins.
Journal of the American Chemical Society (2004-08-26)
A James Link, Mandy K S Vink, David A Tirrell
ABSTRACT
An improved protocol for copper-catalyzed triazole formation on the bacterial cell surface is described. Addition of highly pure CuBr to cells treated with azidohomoalanine (2) leads to ca. 10-fold more extensive cell surface labeling than previously observed. This highly active catalyst allows detection of the methionine analogues azidoalanine (1), azidonorvaline (3), and azidonorleucine (4) in cell surface proteins. Azidoalanine was previously believed to be silent with regard to the cellular protein synthesis machinery.
Sigma-Aldrich
DL-Norvaline
Sigma-Aldrich
L-Norvaline, arginase inhibitor