MilliporeSigma
Inhibition of phosphatidylinositol kinase by toyocamycin.
The Journal of antibiotics (1990-12-01)
H Nishioka, T Sawa, M Hamada, N Shimura, M Imoto, K Umezawa
ABSTRACT
We have screened toyocamycin as an inhibitor of phosphatidylinositol kinase. It inhibited the enzyme of A431 cell membrane with an IC50 of 3.3 micrograms/ml. Adenosine and formycin A also inhibited the enzyme, but other 6 related nucleosides did not. Although orobol and 2,3-dihydroxybenzaldehyde that inhibit phosphatidylinositol kinase inhibited in situ phosphatidylinositol turnover, toyocamycin did not.
Sigma-Aldrich
2,3-Dihydroxybenzaldehyde, 97%