MilliporeSigma
  • Supplemental effects of probiotic Bacillus subtilis fmbJ on growth performance, antioxidant capacity, and meat quality of broiler chickens.
Supplemental effects of probiotic Bacillus subtilis fmbJ on growth performance, antioxidant capacity, and meat quality of broiler chickens.
Poultry science (2016-08-04)
Kaiwen Bai, Qiang Huang, Jingfei Zhang, Jintian He, Lili Zhang, Tian Wang
ABSTRACT
This study aimed to investigate the supplemental effects of probiotic Bacillus subtilis fmbJ (BS fmbJ) on growth performance, antioxidant capacity, and meat quality of broiler chickens. A total of 240 day-old male Arbor Acres (AA) broiler chickens were randomly allotted to 4 treatments and raised for 6 wk. Each treatment had 6 replicate pens with ten birds per replicate. Birds in the control group (CON) were fed diets without BS fmbJ and antibiotics. The BS groups were fed the basal diets with BS fmbJ at 2 × 10
Sigma-Aldrich
Calcium iodate, 98%