MilliporeSigma
  • Wingless ligand 5a is a critical regulator of placental growth and survival.
Wingless ligand 5a is a critical regulator of placental growth and survival.
Scientific reports (2016-06-18)
Gudrun Meinhardt, Leila Saleh, Gerlinde R Otti, Sandra Haider, Philipp Velicky, Christian Fiala, Jürgen Pollheimer, Martin Knöfler
ABSTRACT
The maternal uterine environment is likely critical for human placental morphogenesis and development of its different trophoblast subtypes. However, factors controlling growth and differentiation of these cells during early gestation remain poorly elucidated. Herein, we provide evidence that the ligand Wnt5a could be a critical regulator of trophoblast proliferation and survival. Immunofluorescence of tissues and western blot analyses of primary cultures revealed abundant Wnt5a expression and secretion from first trimester decidual and villous stromal cells. The ligand was also detectable in decidual glands, macrophages and NK cells. Wnt5a increased proliferation of villous cytotrophoblasts and cell column trophoblasts, outgrowth on collagen I as well as cyclin A and D1 expression in floating explant cultures, but suppressed camptothecin-induced apoptosis. Similarly, Wnt5a stimulated BrdU incorporation and decreased caspase-cleaved cytokeratin 18 neo-epitope expression in primary cytotrophoblasts. Moreover, Wnt5a promoted activation of the MAPK pathway in the different trophoblast models. Chemical inhibition of p42/44 MAPK abolished cyclin D1 expression and Wnt5a-stimulated proliferation. Compared to controls, MAPK phosphorylation and proliferation of cytotrophoblasts declined upon supplementation of supernatants from Wnt5a gene-silenced decidual or villous stromal cells. In summary, non-canonical Wnt5a signalling could play a role in early human trophoblast development by promoting cell proliferation and survival.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder