MilliporeSigma
  • Assay to measure sphingomyelinase and ceramidase activities efficiently and safely.
Assay to measure sphingomyelinase and ceramidase activities efficiently and safely.
Journal of chromatography. A (2016-12-26)
Christiane Mühle, Johannes Kornhuber
ABSTRACT
As part of the sphingomyelin pathway, sphingomyelinases and ceramidases have attracted much attention in basic as well as clinical research. However, current assays still often rely on a radioactive substrate, extensive manual purification steps, and hazardous solvents for chromatographic analysis. We here show the equivalence of a fluorescent sphingomyelin substrate and present a new versatile solvent replacing the chloroform/methanol mixture. By further modifications including the omission of the manual extraction steps, chloroform and methanol are eliminated from the entire procedure and render the assay flexible to repeated analyses at multiple time intervals. These improvements allow for the rapid detection of both enzymes in a high throughput microtiter format. Moreover, we demonstrate the relevance of the plastic assay material and the interchangeability between serum and different plasma sources.
Product Description
Supelco
Rectangular TLC developing tanks, complete, L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Supelco
PTFE multi-plate racks, For use with 20 cm × 20 cm plates