MilliporeSigma
  • Evaluation of a selected lactic acid bacteria-based probiotic on Salmonella enterica serovar Enteritidis colonization and intestinal permeability in broiler chickens.
Evaluation of a selected lactic acid bacteria-based probiotic on Salmonella enterica serovar Enteritidis colonization and intestinal permeability in broiler chickens.
Avian pathology : journal of the W.V.P.A (2016-08-23)
Omar F Prado-Rebolledo, Jaime de Jesus Delgado-Machuca, Rafael J Macedo-Barragan, Luis J Garcia-Márquez, Jesus E Morales-Barrera, Juan D Latorre, Xochitl Hernandez-Velasco, Guillermo Tellez
ABSTRACT
Two experiments were conducted to evaluate the effect of a lactic acid bacteria-based probiotic (FloraMax-B11®) against Salmonella enterica serovar Enteritidis intestinal colonization and intestinal permeability in broiler chickens. Experiment 1 consisted of two independent trials. In each trial, day-old broiler chicks were assigned to one of two groups: control + S. Enteritidis or probiotic + S. Enteritidis. At 72 h post-S. Enteritidis challenge, haematology and caecal content were evaluated for S. Enteritidis colonization. In Experiment 2, day-old broiler chicks were assigned to one of four groups: negative control; probiotic; control + S. Enteritidis; or probiotic + S. Enteritidis. At 72 h post-S. Enteritidis challenge, chickens in all groups were given an oral gavage dose of fluorescein isothiocyanate dextran (FITC-d). In both trials of Experiment 1, a significant reduction (P < 0.05) in colony-forming units/gram of S. Enteritidis in caecal content and a reduction in the incidence of S. Enteritidis enriched caecal samples were observed in probiotic + S. Enteritidis chickens. In addition, significant heterophilia and lymphopaenia were observed in control + S. Enteritidis chickens. In Experiment 2, a decrease in numbers of S. Enteritidis in caeca were observed in probiotic + S. Enteritidis chickens when compared to control + S. Enteritidis. Also, an increase in serum FITC-d concentration was detected in control + S. Enteritidis. These results suggest that early infection with S. Enteritidis can increase intestinal permeability, but the adverse effects can be prevented by the administration of the probiotic tested.
Product Description
Millipore
Brilliant green agar, modified, suitable for microbiology, NutriSelect® Basic
Sigma-Aldrich
Nalidixic acid sodium salt, powder
Millipore
Tryptic Soy Broth - Dehydrated Culture Media, NutriSelect® Plus, powder, suitable for microbiology