MilliporeSigma
  • Nanoconfined self-assembly on a grafted graphitic surface under electrochemical control.
Nanoconfined self-assembly on a grafted graphitic surface under electrochemical control.
Nanoscale (2016-12-08)
Thi Mien Trung Huynh, Thanh Hai Phan, Oleksandr Ivasenko, Stijn F L Mertens, Steven De Feyter
ABSTRACT
Highly oriented pyrolytic graphite (HOPG) can be covalently grafted with aryl radicals generated via the electrochemical reduction of 3,5-bis-tert-butyl-diazonium cations (3,5-TBD). The structure of the grafted layer and its stability under electrochemical conditions were assessed with electrochemical scanning tunneling microscopy (EC-STM) and cyclic voltammetry (CV). Stable within a wide (>2.5 V) electrochemical window, the grafted species can be locally removed using EC-STM-tip nanolithography. Using dibenzyl viologen as an example, we show that the generated nanocorrals of bare graphitic surface can be used to study nucleation and growth of self-assembled structures under conditions of nanoconfinement and electrochemical potential control.