MilliporeSigma
  • Novel electrospun nanofibers of modified gelatin-tyrosine in cartilage tissue engineering.
Novel electrospun nanofibers of modified gelatin-tyrosine in cartilage tissue engineering.
Materials science & engineering. C, Materials for biological applications (2016-12-19)
Maria Agheb, Mohammad Dinari, Mohammad Rafienia, Hossein Salehi
ABSTRACT
In natural cartilage tissues, chondrocytes are linked to extracellular matrix (ECM) through cell-surface binding proteins. Surface modification of gelatin can provide a new generation of biopolymers and fibrous scaffolds with chemical, mechanical, and biological properties. In this study tyrosine protein and 1,2,3-triazole ring were utilized to functionalize gelatin without Cu catalyst. Their molecular structure was characterized by Fourier transform infrared spectroscopy (FTIR) and nuclear magnetic resonance spectroscopy (
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
L-Tyrosine methyl ester hydrochloride