MilliporeSigma
  • Lunasin is a novel therapeutic agent for targeting melanoma cancer stem cells.
Lunasin is a novel therapeutic agent for targeting melanoma cancer stem cells.
Oncotarget (2016-08-28)
Chris Shidal, Numan Al-Rayyan, Kavitha Yaddanapudi, Keith R Davis
ABSTRACT
Recent studies provide compelling evidence that melanoma is initiated and maintained by a small population of malignant cells called cancer-initiating cells (CICs) that exhibit stem-cell-like properties. Observations that CICs have a distinct biology when compared to that of the bulk tumor cells and, importantly, are resistant to chemotherapies and radiation, suggest that CICs are involved in invasion, metastasis, and ultimately relapse. Lunasin, a bioactive peptide present in soybean, has both chemopreventive activity and chemotherapeutic activity against multiple cancer types. In this study, we tested the potential of Lunasin to specifically target CICs in melanoma tumor cell populations. In vitro studies using human melanoma cell lines showed that Lunasin treatment decreased the size of a subpopulation of melanoma cells expressing the surrogate CIC marker, Aldehyde Dehydrogenase, concomitant with a reduction in the ability to form colonies in soft agar assays, and reduced tumor growth in mouse xenografts. Similarly, Lunasin inhibited colony formation by isolated melanoma CICs in soft agar and reduced oncosphere formation in vitro and substantially inhibited tumor growth in mouse xenografts. Mechanistic studies revealed that Lunasin treatment of isolated melanoma CICs induced expression of the melanocyte-associated differentiation markers Tyrosinase and Microphthalmia-associated Transcription Factor concomitant with reduced expression of the stemness factor NANOG. These findings document for the first time that Lunasin has significant therapeutic activity against melanoma by specifically targeting melanoma CICs, and inducing a more differentiated, non-CIC phenotype. Thus, Lunasin may represent a novel therapeutic option for both chemoresistant and advanced metastatic melanoma management.
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Anti-NANOG Antibody, clone 7F7.1, clone 7F7.1, from mouse
Sigma-Aldrich
Anti-Tyrosinase Antibody, clone T311, clone T311, Upstate®, from mouse